L-(+)-Pantolactone 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Hydroxy-4,4-dimethyldihydrofuran-2(3H)-one

A206774-100MG A206774-1G A206774-250MG A206774-5G
103848-410EA 136 USD
103848-410 77164-810 77164-812 77164-814 77303-008
L-(+)-Pantolactone 97%
L-(+)-Pantolactone

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.143 g/mol
Boiling Pt: 224.6 °C (760 mm Hg)
Melting Pt: 90…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216625
CAS Number: 5405-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR