(S)-(-)-3-Amino-1,2-propanediol 97%
Supplier: Ambeed
|
Danger
|
A107298-10G
A107298-25G
A107298-1G
A107298-5G
A107298-100G
103797-162EA
5
USD
103797-162
103822-930
76809-744
76809-746
77309-780
77309-782
77882-905
(S)-(-)-3-Amino-1,2-propanediol 97%
(S)-(-)-3-Amino-1,2-propanediol
Precaución: for Research Use Only
Formula:
C₃H₉NO₂ Boiling Pt: 266.7 °C (760 mm Hg) Melting Pt: 55 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00798260 CAS Number: 61278-21-5 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...