(S)-3-Aminopiperidin-2-one hydrochloride 97%

Supplier: Ambeed
Warning

A235943-250MG A235943-1G
103811-052EA 68 USD
103811-052 103850-822 77193-216 77304-026
(S)-3-Aminopiperidin-2-one hydrochloride 97%
(S)-3-Aminopiperidin-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁ClN₂O
MW: 150.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09259964
CAS Number: 42538-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR