(S)-3-Aminohexan-1-ol hydrochloride 97%

Supplier: Ambeed
Warning

A210536-100MG A210536-250MG A210536-5G A210536-1G
103818-012EA 131 USD
103818-012 76825-866 76825-868 76825-870 77087-082
(S)-3-Aminohexan-1-ol hydrochloride 97%
(S)-3-Aminohexan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆ClNO
MW: 153.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701471
CAS Number: 68889-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR