(S)-3-Aminobutan-1-ol 95%

Supplier: Ambeed
Danger

A104138-100MG A104138-250MG A104138-25G A104138-1G A104138-5G
103796-658EA 16 USD
103796-658 103822-372 76970-314 76970-316 76970-318 77309-234 77309-236
(S)-3-Aminobutan-1-ol 95%
(S)-3-Aminobutan-1-ol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MW: 89.13 g/mol
Boiling Pt: 168 °C (760 mm Hg)
Flash Pt: 55.616 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275763
CAS Number: 61477-39-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR