(S)-3-Amino-4-(naphthalen-1-yl)butanoic acid hydrochloride 98%

Supplier: Ambeed
Warning

A134587-100MG A134587-1G
103802-024EA 171 USD
103802-024 76782-580 77314-788
(S)-3-Amino-4-(naphthalen-1-yl)butanoic acid hydrochloride 98%
(S)-3-Amino-4-(naphthalen-1-yl)butanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₆ClNO₂
MW: 265.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861033
CAS Number: 331846-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR