O-Acetyl-L-serine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Acetoxy-2-aminopropanoic acid hydrochloride, H-Ser(Ac)-OH.HCl

A266030-10G A266030-250MG A266030-1G
103812-776EA 23 USD
103812-776 103852-672 76722-942 76722-944 77092-370
O-Acetyl-L-serine hydrochloride 95%
O-Acetyl-L-serine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉NO₄
MW: 183.59 g/mol
Melting Pt: 163 °C
Storage Temperature: Freezer
MDL Number: MFCD00060169
CAS Number: 66638-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR