(S)-3,3-Dimethyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one 95%

Supplier: Ambeed
Warning

A170325-250MG A170325-1G A170325-5G
103806-470EA 162 USD
103806-470 103844-744 76985-378 77301-038 77301-040
(S)-3,3-Dimethyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one 95%
(S)-3,3-Dimethyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MW: 155.19 g/mol
Boiling Pt: 262.2±19.0 °C (760 mm Hg)
Melting Pt: 38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD26940310
CAS Number: 99208-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR