(S)-(-)-3-tert-Butylamino-1,2-propanediol 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(tert-Butylamino)propane-1,2-diol

A244050-100G A244050-25G A244050-5G
103851-392EA 10 USD
103851-392 76771-480 77090-596 77090-598
(S)-(-)-3-tert-Butylamino-1,2-propanediol 98%
(S)-(-)-3-tert-Butylamino-1,2-propanediol

Precaución: for Research Use Only

Formula: C₇H₁₇NO₂
MW: 147.22 g/mol
Boiling Pt: 262.4 °C (760 mm Hg)
Melting Pt: 85…89 °C
Flash Pt: 109.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00190165
CAS Number: 30315-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR