(S)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-3-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

A310952-250MG A310952-5G A310952-1G A310952-10G A310952-25G
103814-640EA 86 USD
103814-640 77002-684 77002-686 77306-040 77672-584 77672-586
(S)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid 95%
(S)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.2010 g/mol
Boiling Pt: 479.1 °C (760 mm Hg)
Melting Pt: 143.8 °C
Flash Pt: 243.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03419292
CAS Number: 261165-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR