(S)-3-(3-(Allyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A133055-100MG
A133055-250MG
A133055-1G
103801-816EA
74
USD
103801-816
76983-830
76983-832
77298-602
(S)-3-(3-(Allyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 95%
(S)-3-(3-(Allyloxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₇H₂₃NO₅ MW: 321.37 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08061618 CAS Number: 1175919-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...