(S)-3-(1-Aminoethyl)benzonitrile hydrochloride 95%

Supplier: Ambeed
Warning

A295631-100MG A295631-250MG A295631-5G
103814-068EA 204 USD
103814-068 76816-652 76816-654 76816-656
(S)-3-(1-Aminoethyl)benzonitrile hydrochloride 95%
(S)-3-(1-Aminoethyl)benzonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11101269
CAS Number: 2741870-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR