(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-Biphenylalanine, H-4-Phenyl-Phe-OH
A324890-100MG
A324890-250MG
A324890-1G
103825-384EA
19
USD
103825-384
103836-326
77263-494
77263-496
77306-324
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid 97%
(S)-3-([1,1'-Biphenyl]-4-yl)-2-aminopropanoic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₅NO₂ MW: 241.29 g/mol Boiling Pt: 569.2 °C (760 mm Hg) Flash Pt: 298.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318878 CAS Number: 155760-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...