(S)-3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-IsoSer-OH

A250483-100MG A250483-250MG
103811-928EA 58 USD
103811-928 103851-768 77031-494 77031-496
(S)-3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid 98%
(S)-3-((tert-Butoxycarbonyl)amino)-2-hydroxypropanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₅NO₅
MW: 205.21 g/mol
Boiling Pt: 391.7 °C (760 mm Hg)
Melting Pt: 83…87 °C
Flash Pt: 190.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17011798
CAS Number: 52558-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR