(S)-3-((2R,5S)-5-(4-Fluorophenyl)-2-((S)-((4-fluorophenyl)amino)(4-((trimethylsilyl)oxy)phenyl)methyl)-5-((trimethylsilyl)oxy)pentanoyl)-4-phenyloxazolidin-2-one 97%

Supplier: Ambeed
Warning

A415517-1G A415517-5G A415517-100G A415517-250MG A415517-25G
103827-296EA 9 USD
103827-296 103838-340 76841-922 76841-924 77004-058 77004-060 77004-062
(S)-3-((2R,5S)-5-(4-Fluorophenyl)-2-((S)-((4-fluorophenyl)amino)(4-((trimethylsilyl)oxy)phenyl)methyl)-5-((trimethylsilyl)oxy)pentanoyl)-4-phenyloxazolidin-2-one 97%
(S)-3-((2R,5S)-5-(4-Fluorophenyl)-2-((S)-((4-fluorophenyl)amino)(4-((trimethylsilyl)oxy)phenyl)methyl)-5-((trimethylsilyl)oxy)pentanoyl)-4-phenyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₃₉H₄₆F₂N₂O₅Si₂
MW: 716.96 g/mol
Boiling Pt: 748.9 °C (760 mm Hg)
Flash Pt: 406.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08460200
CAS Number: 272778-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR