Fmoc-D-β-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: FMOC-D-beta-phenylalanine, (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid, (S)-N-FMOC-3-phenyl-3-alanine, (S)-N-(9-Fluorenylmethoxycarbonyl)-3-amino-3-phenylpropanoic acid, Fmoc-D-β-Phe-OH

A135142-250MG A135142-25G A135142-1G A135142-5G
103802-096EA 19 USD
103802-096 76996-596 76996-598 77314-886 77314-888
Fmoc-D-β-phenylalanine 95%
Fmoc-D-beta-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.44 g/mol
Boiling Pt: 617 °C (760 mm Hg)
Melting Pt: 187.3 °C
Flash Pt: 326.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311759
CAS Number: 209252-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR