(S)-Fmoc-β2-homoalanine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-methylpropanoic acid, N,(S)-Fmoc-β2-homoalanine

A280622-100MG A280622-250MG
103813-516EA 185 USD
103813-516 77099-166 77099-168
(S)-Fmoc-β2-homoalanine 98%
(S)-Fmoc-β2-homoalanine

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.358 g/mol
Boiling Pt: 555.3 °C (760 mm Hg)
Flash Pt: 289.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03788584
CAS Number: 203854-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR