(S)-(+)-Hydratropic acid 98%

Supplier: Ambeed
Warning

A117720-100G A117720-10G A117720-25G A117720-1G A117720-5G
103798-928EA 9 USD
103798-928 103824-818 76687-964 76687-966 76687-968 77311-596 77311-598
(S)-(+)-Hydratropic acid 98%
(S)-(+)-Hydratropic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
Boiling Pt: 261.0±0.0 °C (760 mm Hg)
Melting Pt: 29…30 °C
Flash Pt: 154.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00063139
CAS Number: 7782-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR