(S)-2-Hydroxybutanoic acid 96%

Supplier: Ambeed
Danger

A186753-100MG A186753-10G A186753-250MG A186753-1G A186753-25G A186753-5G
103815-960EA 12 USD
103815-960 103846-548 76974-678 76974-680 76974-682 77084-232 77084-234 77084-236
(S)-2-Hydroxybutanoic acid 96%
(S)-2-Hydroxybutanoic acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.1045 g/mol
Boiling Pt: 238.3 °C (760 mm Hg)
Melting Pt: 50…54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318568
CAS Number: 3347-90-8
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR