(S)-(+)-2-Chloro-1-propanol 98%

Supplier: Ambeed
Danger

Synonyms: (S)-2-Chloropropan-1-ol

A659605-250MG A659605-1G
103840-434EA 217 USD
103840-434 77145-362 77308-074
(S)-(+)-2-Chloro-1-propanol 98%
(S)-(+)-2-Chloro-1-propanol

Precaución: for Research Use Only

Formula: H₃CCHClCH₂OH
MW: 94.54 g/mol
Boiling Pt: 133.5 °C (760 mm Hg)
Flash Pt: 44.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010812
CAS Number: 19210-21-0
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR