L-Propargylglycine 97%

Supplier: Ambeed
Warning

Synonyms: H-Pra-OH, (S)-2-Aminopent-4-ynoic acid, (S)-alpha-Propargylglycine

A129076-10G A129076-250MG A129076-1G A129076-5G
103801-222EA 36 USD
103801-222 76996-248 76996-250 77313-588 77313-590
L-Propargylglycine 97%
L-Propargylglycine

Precaución: for Research Use Only

Formula: C₅H₇NO₂
Boiling Pt: 272 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077855
CAS Number: 23235-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR