(S)-2-Aminooctanoic acid 97%

Supplier: Ambeed
Warning

A109740-10G A109740-250MG A109740-25G A109740-1G A109740-5G
103797-556EA 19 USD
103797-556 103823-362 77148-652 77148-654 77148-656 77310-172 77310-174
(S)-2-Aminooctanoic acid 97%
(S)-2-Aminooctanoic acid

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂
MW: 159.2300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03701463
CAS Number: 116783-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR