(S)-2-Aminoheptanoic acid 98%

Supplier: Ambeed
Warning

A322366-5G A322366-250MG A322366-1G
103825-304EA 20 USD
103825-304 103836-238 76839-158 77099-874 77306-264
(S)-2-Aminoheptanoic acid 98%
(S)-2-Aminoheptanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.19 g/mol
Boiling Pt: 251 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08275754
CAS Number: 44902-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR