(S)-(+)-2-Amino-1-butanol 98%

Supplier: Ambeed
Danger

Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol

A424986-10G A424986-100G
103838-502EA 6 USD
103838-502 76818-060 77673-036
(S)-(+)-2-Amino-1-butanol 98%
(S)-(+)-2-Amino-1-butanol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 177.2±13.0 °C (760 mm Hg)
Melting Pt: 2 °C
Flash Pt: 82.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064418
CAS Number: 5856-62-2
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR