(S)-2-Aminobut-3-en-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

A296061-1G A296061-5G A296061-250MG
103814-084EA 78 USD
103814-084 103835-218 76837-882 76837-884 77114-384
(S)-2-Aminobut-3-en-1-ol hydrochloride 95%
(S)-2-Aminobut-3-en-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
Boiling Pt: 174.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09025316
CAS Number: 219803-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR