Symmetric dimethylarginine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
NG,NG'-Dimethyl-L-arginine, (S)-2-Amino-5-((bis(methylamino)methylene)amino)pentanoic acid, N5-[(Methylamino)(methylimino)methyl]-L-ornithine, SDMA
A507261-10MG
A507261-1MG
A507261-25MG
A507261-100MG
A507261-250MG
103792-750EA
93
USD
103792-750
103799-668
103828-344
77103-058
77103-060
77103-062
77613-600
77686-324
Symmetric dimethylarginine 98%
Symmetric dimethylarginine
Precaución: for Research Use Only
Formula:
C₈H₁₈N₄O₂ MW: 202.25 g/mol Boiling Pt: 367.7 °C (760 mm Hg) Flash Pt: 176.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD14705169 CAS Number: 30344-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...