γ-Methyl-L-leucine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-β-tBu-Ala-OH, (S)-2-Amino-4,4-dimethylpentanoic acid, H-4-Me-Leu-OH
A218673-100MG
A218673-1G
A218673-5G
A218673-250MG
A218673-25G
103818-728EA
123
USD
103818-728
103849-598
76800-828
77088-126
77303-500
77879-336
77879-337
γ-Methyl-L-leucine 95%
gamma-Methyl-L-leucine
Precaución: for Research Use Only
Formula:
C₇H₁₅NO₂ MW: 145.202 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00066079 CAS Number: 57224-50-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...