L-Cyclopropylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-cyclopropylpropanoic acid, H-Cyclopropyl-Ala-OH, H-β-Cyclopropyl-Ala-OH

A132363-10G A132363-250MG A132363-1G A132363-25G A132363-5G A132363-100G
103801-728EA 18 USD
103801-728 77285-368 77285-370 77314-304 77314-306 77314-308 77615-052
L-Cyclopropylalanine 97%
L-Cyclopropylalanine

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.159 g/mol
Boiling Pt: 258 °C (760 mm Hg)
Flash Pt: 109.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00798687
CAS Number: 102735-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR