(S)-2-Amino-3-cyclohexylpropanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Cyclohexyl-L-alanine

A179780-1G A179780-25G A179780-5G A179780-100G A179780-10G A179780-500G
103807-462EA 9 USD
103807-462 103845-814 77083-286 77083-288 77083-290 77260-184 77260-186 77260-188
(S)-2-Amino-3-cyclohexylpropanoic acid 97%
(S)-2-Amino-3-cyclohexylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.24 g/mol
Melting Pt: 322 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153484
CAS Number: 27527-05-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR