3,3-Diphenyl-L-alanine 97%

Supplier: Ambeed
Warning

Synonyms: H-Ala(3,3-DiPhenyl)-OH, (S)-2-Amino-3,3-diphenylpropanoic acid

A277050-10G A277050-250MG A277050-1G A277050-25G A277050-5G A277050-100G
103813-330EA 29 USD
103813-330 103853-266 77002-086 77002-088 77093-140 77093-142 77093-144 77672-426
3,3-Diphenyl-L-alanine 97%
3,3-Diphenyl-L-alanine

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₂
MW: 241.29 g/mol
Boiling Pt: 389.2 °C (760 mm Hg)
Flash Pt: 189.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01631989
CAS Number: 149597-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR