7-Chloro-L-tryptophan 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(7-chloro-1H-indol-3-yl)propanoic acid

A175659-1G A175659-100MG A175659-250MG
103807-074EA 377 USD
103807-074 77082-728 77163-700 77163-702
7-Chloro-L-tryptophan 97%
7-Chloro-L-tryptophan

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
Boiling Pt: 476.9±45.0 °C (760 mm Hg)
Melting Pt: 290…291 °C
Flash Pt: 242.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09264322
CAS Number: 73945-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR