5-Fluoro-L-tryptophan 98%

Supplier: Ambeed
Warning

A370516-1G A370516-250MG A370516-100MG A370516-5G
103826-462EA 100 USD
103826-462 76840-722 77195-490 77616-634 77746-747
5-Fluoro-L-tryptophan 98%
5-Fluoro-L-tryptophan

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂O₂
Boiling Pt: 450.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00066470
CAS Number: 16626-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR