(S)-2-Amino-3-(4-fluorophenyl)propanoic acid hydrochloride 97%

Supplier: Ambeed
Warning

A418880-1G A418880-10G
103827-362EA 15 USD
103827-362 103838-406 76842-018 77291-378
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid hydrochloride 97%
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClFNO₂
Melting Pt: 245…251 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863499
CAS Number: 64231-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR