4-Fluoro-L-phenylalanine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH
A183300-100G
A183300-10G
A183300-1G
A183300-25G
A183300-5G
103815-628EA
6
USD
103815-628
103846-190
76690-630
76690-632
77083-790
77083-792
77083-794
4-Fluoro-L-phenylalanine 98%
4-Fluoro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₉H₁₀FNO₂ MW: 183.2 g/mol Melting Pt: 245…251 °C Flash Pt: 161.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063064 CAS Number: 1132-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...