4-Borono-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: H-Phe(4-B)-OH, L-4-(2-Amino-2-carboxyethyl)benzeneboronic acid, (S)-2-Amino-3-(4-boronophenyl)propanoic acid

A134259-100MG A134259-1G A134259-5G A134259-25G
103801-992EA 59 USD
103801-992 77246-666 77298-704 77314-728 77745-955
4-Borono-L-phenylalanine 95%
4-Borono-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₂BNO₄
MW: 209.008 g/mol
Melting Pt: >290 °C
Flash Pt: 225.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075172
CAS Number: 76410-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR