3-Fluoro-L-tyrosine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-hydroxy-L-phenylalanine, (S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid, H-Tyr(3-F)-OH

A374537-1G A374537-100MG A374537-250MG
103826-554EA 163 USD
103826-554 76840-852 77251-816 77251-818
3-Fluoro-L-tyrosine 97%
3-Fluoro-L-tyrosine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002607
CAS Number: 7423-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR