3,4-Difluoro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(3,4-difluorophenyl)propanoic acid, H-Phe(3,4-DiF)-OH

A101708-100MG A101708-25G A101708-1G A101708-5G
103796-100EA 34 USD
103796-100 103821-788 76994-530 76994-532 77282-810 77282-812
3,4-Difluoro-L-phenylalanine 97%
3,4-Difluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.18 g/mol
Boiling Pt: 311.9 °C (760 mm Hg)
Flash Pt: 142.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01075125
CAS Number: 31105-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR