2,4-Difluoro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-3-(2,4-difluorophenyl)propanoic acid

A516581-250MG A516581-1G A516581-5G
103828-456EA 55 USD
103828-456 103839-562 77004-816 77307-676 77307-678
2,4-Difluoro-L-phenylalanine 97%
2,4-Difluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₉F₂NO₂
MW: 201.17 g/mol
Boiling Pt: 306.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03844652
CAS Number: 31105-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR