(S)-2-Cyclobutylglycine 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-2-cyclobutylacetic acid

A362585-100MG A362585-250MG
103826-326EA 82 USD
103826-326 76978-602 76978-604
(S)-2-Cyclobutylglycine 97%
(S)-2-Cyclobutylglycine

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
MW: 129.157 g/mol
Boiling Pt: 261 °C (760 mm Hg)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD08436202
CAS Number: 49607-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR