(S)-2-amino-2,3-dimethylbutanoic acid 95%
Supplier: Ambeed
|
Warning
|
A150369-100MG
A150369-10G
A150369-250MG
A150369-1G
103804-152EA
155
USD
103804-152
103842-286
77150-358
77150-360
77150-362
77317-802
(S)-2-amino-2,3-dimethylbutanoic acid 95%
(S)-2-amino-2,3-dimethylbutanoic acid
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ Boiling Pt: 217.7 °C (760 mm Hg) Flash Pt: 85.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318764 CAS Number: 53940-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...