(S)-2-Amino-2-(3-chlorophenyl)ethanol 97%
Supplier: Ambeed
|
Warning
|
A148014-10G
A148014-250MG
A148014-1G
A148014-25G
A148014-5G
103803-868EA
40
USD
103803-868
103841-984
76871-498
76871-500
77317-416
77317-418
77317-420
(S)-2-Amino-2-(3-chlorophenyl)ethanol 97%
(S)-2-Amino-2-(3-chlorophenyl)ethanol
Precaución: for Research Use Only
Formula:
C₈H₁₀ClNO MW: 171.624 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09253677 CAS Number: 663611-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...