L-(+)-2-(2-Chlorophenyl)glycine ≥95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-2-(2-chlorophenyl)acetic acid, (S)-(+)-2-Amino-2-(2-chlorophenyl)acetic Acid, H-Phg(2-Cl)-OH, 2-Chloro-L-(+)-2-phenylglycine

A385418-10G A385418-100G
103837-818EA 29 USD
103837-818 77878-117 77878-116
L-(+)-2-(2-Chlorophenyl)glycine ≥95%
L-(+)-2-(2-Chlorophenyl)glycine

Precaución: Keep in a dark place, inert atmosphere, room temperature

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 386.6 °C (760 mm Hg)
Melting Pt: 182…187 °C
Flash Pt: 187.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03426346
CAS Number: 141315-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR