Ac-Asp(OtBu)-OH 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Acetyl-L-aspartic acid 4-tert-butyl ester, (S)-2-Acetamido-4-(tert-butoxy)-4-oxobutanoic acid, N-Ac-Asp(OtBu)-OH
A121743-25G
A121743-100G
A121743-10G
103800-128EA
14
USD
103800-128
77566-262
77624-010
77876-881
Ac-Asp(OtBu)-OH 98%
Ac-Asp(OtBu)-OH
Precaución: For research use only. Sealed in dry, at 2…8°C
Formula:
C₁₀H₁₇NO₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD00236746 CAS Number: 117833-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...