(S)-2,4-Diaminobutanoic acid dihydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-2,4-Diaminobutanoic acid dihydrochloride, H-Dab-OH 2HCl, L-2,4-Diaminobutyric acid dihydrochloride, (2S)-2,4-Diaminobutanoic acid dihydrochloride, H-Daba-OH·2HCl, (S)-(+)-2,4-Diaminobutyric acid dihydrochloride
A217984-25G
A217984-1G
103818-680EA
6
USD
103818-680
103849-412
77088-064
77303-472
(S)-2,4-Diaminobutanoic acid dihydrochloride 95%
(S)-2,4-Diaminobutanoic acid dihydrochloride
Precaución: for Research Use Only
Formula:
C₄H₁₂Cl₂N₂O₂ MW: 191.06 g/mol Boiling Pt: 321 °C (760 mm Hg) Melting Pt: 191…195 °C Flash Pt: 147.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064561 CAS Number: 1883-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...