(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: Ambeed
Warning

A376699-100G A376699-100MG A376699-10G A376699-250MG A376699-1G A376699-25G A376699-5G
103826-588EA 9 USD
103826-588 103837-614 77290-906 77290-908 77290-910 77290-912 77306-884 77306-886 77306-888
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(S)-(-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₄₄H₃₂P₂
Boiling Pt: 724.3 °C (760 mm Hg)
Melting Pt: 283…286 °C
Flash Pt: 419 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR