(S)-2-(Piperidin-2-yl)ethanol 97%

Supplier: Ambeed
Warning

A199536-100MG A199536-10G A199536-250MG A199536-1G
103817-112EA 50 USD
103817-112 103847-736 76975-188 76975-190 76975-192 77085-778
(S)-2-(Piperidin-2-yl)ethanol 97%
(S)-2-(Piperidin-2-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₁₅NO
Melting Pt: 60…62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08461072
CAS Number: 103639-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR