(S)-2-(Methoxymethyl)-morpholine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-(Methoxymethyl)morpholine hydrochloride
A184380-100MG
A184380-250MG
A184380-1G
103815-750EA
99
USD
103815-750
103846-312
76825-046
76825-048
77083-924
(S)-2-(Methoxymethyl)-morpholine hydrochloride 95%
(S)-2-(Methoxymethyl)-morpholine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂·ClH MW: 167.634 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294688 CAS Number: 141196-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...