(S)-2-(Adamantan-1-yl)-2-(((R)-2-hydroxy-1-phenylethyl)amino)acetic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A221372-1G
A221372-10G
A221372-250MG
103818-918EA
9
USD
103818-918
103849-798
77088-394
77165-220
77165-222
(S)-2-(Adamantan-1-yl)-2-(((R)-2-hydroxy-1-phenylethyl)amino)acetic acid hydrochloride 97%
(S)-2-(Adamantan-1-yl)-2-(((R)-2-hydroxy-1-phenylethyl)amino)acetic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₂₀H₂₈ClNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD28016457 CAS Number: 361441-96-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...