(S)-4-Chloromandelic acid 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(4-Chlorophenyl)-2-hydroxyacetic acid

A121286-100MG A121286-250MG A121286-1G A121286-5G
103800-056EA 54 USD
103800-056 77189-684 77189-686 77312-174 77312-176
(S)-4-Chloromandelic acid 98%
(S)-4-Chloromandelic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00798439
CAS Number: 76496-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR