(S)-2-(4-Aminobenzamido)pentanedioic acid 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-Aminobenzoyl)-L-glutamic acid, N-(4-NH₂-Bz)-Glu-OH

A133828-100G A133828-10G A133828-1G A133828-25G A133828-5G
76972-242EA 21.22 USD
76972-242 76972-244 76972-246 77314-614 77314-616
(S)-2-(4-Aminobenzamido)pentanedioic acid 97%
(S)-2-(4-Aminobenzamido)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₅
MW: 266.2500 g/mol
Boiling Pt: 607.1 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 320.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00042821
CAS Number: 4271-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR